missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecanoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Heptadecanoyl Chloride |
| CAS | 40480-10-2 |
| Molecular Formula | C17H33ClO |
| Quantity | 250 mg |
| Synonym | Heptadecanoic Acid Chloride, n-Heptadecanoyl Chloride |
| InChI Formula | InChI=1S/C17H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3 |
| SMILES | ClC(CCCCCCCCCCCCCCCC)=O |
| IUPAC Name | heptadecanoyl chloride |
| Molecular Weight (g/mol) | 288.9 |
| Formula Weight | 288.22 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?