missing translation for 'onlineSavingsMsg'
Learn More

Hepsulfam, MedChemExpress

Product Code. 16470676
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470676 20 mg 20mg
16470159 10 mg 10mg
16442358 5 mg 5mg
16413149 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470676 Supplier Medchem Express Supplier No. HYU00095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hepsulfam (NCI 329680; ZINC01574758) is a anticancer agent that shows excellent antileukemic activity with an median IC50 of 0.91 μg/mL in a panel of different tumors.

Hepsulfam (NCI 329680

Specifications

Chemical Name or Material Hepsulfam
CAS 96892-57-8
Color White
Purity Grade Notes Research
Molecular Formula C7H18N2O6S2
Quantity 20 mg
Synonym NCI 329680 ZINC01574758
SMILES O=S(OCCCCCCCOS(=O)(N)=O)(N)=O
Molecular Weight (g/mol) 290.36
Formula Weight 290.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.