missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepes, TRC
Specifications
Specifications
| Chemical Name or Material | HEPES |
| CAS | 7365-45-9 |
| Molecular Formula | C8 H18 N2 O4 S |
| Quantity | 500 mg |
| Synonym | 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, HEPES, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid), 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid, 1-[4-(2-Hydroxyethyl)-1-piperazinyl]ethane-2-sulfonic acid, 2-[4-(2-Hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid, 2-[4-(2-Hydroxyethyl)piperazin-1-yl]ethanesulfonic acid, 2-[4-(2-Hydroxyethyl)piperazinyl]ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazine-1-(2-ethanesulfonic acid), N-(2-Hydroxyethyl)piperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-2'-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-ethanesulfonic acid, N-2-Hydroxyethylpiperazino-N'-2-ethanesulfonic acid, NSC 166663, N'-(2-Hydroxyethyl)piperazine-N-(2-ethanesulfonic acid), N'-2-Hydroxyethylpiperazine-N-2-ethanesulfonic acid, TVZ 7, WAS 13 |
| InChI Formula | InChI=1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14) |
| SMILES | OCCN1CCN(CCS(=O)(=O)O)CC1 |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
| Molecular Weight (g/mol) | 238.3 |
| Formula Weight | 238.0987 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?