missing translation for 'onlineSavingsMsg'
Learn More

HEPES Sodium Salt, Cell Culture Reagent Grade, ≥98.0%, MP Biomedicals™

Product Code. 11470832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470832 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470832 Supplier MP Biomedicals Supplier No. 0219455010
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity HEPES sodium salt, optimized for cell culture, media formulations, and protein research applications.

Specifications

CAS 75277-39-3
Color White
pH 9.5 - 10.8(1% aq soln)
Physical Form Powder
Quantity 10 g
Solution Type Buffer
Assay Percent Range ≥98.0%
Molecular Formula C8H17N2NaO4S
MDL Number MFCD00036463
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N-(2-ethanesulfonic acid) sodium salt
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Weight (g/mol) 260.28
PubChem CID 2724248
ChEBI CHEBI:46758
Formula Weight 260.28
Percent Purity ≥98%
Grade Cell Culture
Chemical Name or Material HEPES Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.