missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES hemisodium salt

Product Code. 15464329
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464329 500 g 500g
15444329 100 g 100g
15454329 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15464329 Supplier Thermo Scientific Chemicals Supplier No. J61830.36
  • ONLINE EXCLUSIVE PRICE
    £325.38
    £272.65 / 500g
    Save £52.73
    16% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A zwitterionic buffer | CAS: 103404-87-1 | C8H17N2NaO4S | 260.28 g/mol

HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-87-1
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Formulation 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic Acid Sodium Salt
Solubility Information Soluble in water.
Formula Weight 497.58
Chemical Name or Material Zwitterionic Buffer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.