missing translation for 'onlineSavingsMsg'
Learn More

HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™
BIOPHARMA

Product Code. 11416561 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416561 25 g 25g
11426561 100 g 100g
11436561 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416561 Supplier MP Biomedicals Supplier No. 0219482725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Buffering agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >99%
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Formula Weight 238.3
Grade Molecular Biology Reagent
Chemical Name or Material HEPES, free acid, Free Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.