missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES-buffered saline, pH 6.5 (5X)

Product Code. 15493849
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493849 500 mL 500mL
15483849 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493849 Supplier Thermo Scientific Chemicals Supplier No. J61239.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility
Fully miscible in water.

Notes
Contains 125mM HEPES and 750mM sodium chloride, pH 6.5
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color Colorless
Concentration 5X
pH 6.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Solubility Information Fully miscible in water.
Chemical Name or Material HEPES-buffered saline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.