missing translation for 'onlineSavingsMsg'
Learn More

HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™

Product Code. 10409893
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409893 1 L Nalgene Poly Bottle 1 L
10041703 100 mL Poly Bottle 100mL
10648593 500 mL Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10409893 Supplier Fisher BioReagents Supplier No. BP2991
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance ≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
Chemical Name or Material HEPES Buffer
Name Note 1M Solution, pH 7.3
CAS 7365-45-9, 7732-18-5
Color Undesignated
Concentration 0.95 to 1.05 M
pH 7.2 to 7.5
Physical Form Liquid
Quantity 1 L
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Identification Pass Test
Molecular Formula C8H18N2O4S
Packaging Nalgene Poly Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Formula Weight 238.197
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.