missing translation for 'onlineSavingsMsg'
Learn More

HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure

Product Code. 15855388 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15855388 500 mL Nalgene bottle 500mL
15624198 100 mL Nalgene bottle 100mL
15865388 1 L Nalgene bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15855388 Supplier Thermo Scientific Supplier No. J16924.AP
  • ONLINE EXCLUSIVE PRICE
    £144.84
    £120.65 / 500mL
    Save £24.19
    16% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1 M
pH 7.3
Physical Form Solution
Quantity 500 mL
Packaging Nalgene bottle
Beilstein 883043
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Grade Molecular Biology
Chemical Name or Material HEPES

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.