missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HEPES, 1M Buffer Solution
GREENER_CHOICE

Product Code. 11570496
Click to view available options
Quantity:
20 mL
20 x 100 mL
Unit Size:
20mL
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11570496

Brand: Gibco™ 15630122

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

HEPES, 1M Buffer Solution

Gibco™ HEPES (N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10 – 25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator. HEPES is a good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable, and has very low visible and UV light absorbance.

Product Use
For in vitro diagnostic use.

Dual-site cGMP Manufacturing and Quality System
Gibco™ HEPES is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HEPES product made in our Grand Island facility (15630-130). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance UV light
Chemical Name or Material HEPES buffer
Certificate of Analysis FDA, ISO 13485 standards
Description HEPES Buffer Solution, 20 x 100mL, Liquid, 1M Concentration, 7.2 to 7.5 pH, 2 to 8°C Storage, 24 Months Shelf Life, Water Soluble, For use with Flow Cytometry, LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays
Concentration 1 M
For Use With (Application) Mammalian cell culture
pH 7.2 to 7.5
Physical Form Liquid
Quantity 20 x 100 mL
Packaging 1 Each
Solubility Information Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.