missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HEPES
GREENER_CHOICE

Product Code. 18074882
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 18074882

Brand: Tocris Bioscience™ 3173/250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Multipurpose buffer

Multi purpose buffer used in biological research.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 g
Molecular Formula C8H18N2O4S
Product Type HEPES
Molecular Weight (g/mol) 238.30
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.