missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Hepcidin Antimicrobial Peptide Antibody (Ludwig-Maximilians U. anti_Hepsin) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28378-1mg
This item is not returnable.
View return policy
Description
Hepcidin Antimicrobial Peptide Monoclonal antibody specifically detects Hepcidin Antimicrobial Peptide in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Hepcidin Antimicrobial Peptide | |
| Monoclonal | |
| Unconjugated | |
| P81172 | |
| Human | |
| Protein A purified | |
| RUO | |
| 57817 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Ludwig-Maximilians U. anti_Hepsin | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2Bhepcidin, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1 | |
| Hepcidin/ HAMP | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction