missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Hepatocyte Wash Medium
GREENER_CHOICE

Product Code. 11560536
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560536

Brand: Gibco™ 17704024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Hepatocyte Wash Medium

Hepatocyte Wash Medium is an enriched Williams' Medium E for the reduction of cell membrane leakage, ease of centrifugation and high viability.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Hepatocyte Wash Medium
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.