missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HepPar1), FITC, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human, Mouse, Canine samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | Hepatocyte Specific Antigen |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | HepPar1 |
| Conjugate | FITC |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence |
| Formulation | PBS |
| Gene Alias | Hepatocyte, HSA |
| Host Species | Mouse |
| Immunogen | Extract of a formalin-fixed, rejected-allograft of a human liver |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?