missing translation for 'onlineSavingsMsg'
Learn More

Hepatocyte Specific Antigen Antibody (HepPar1), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18353762 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353762 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353762 Supplier Novus Biologicals Supplier No. NBP308970AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatocyte Specific Antigen
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone HepPar1
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Hepatocyte, HSA
Host Species Mouse
Immunogen Extract of a formalin-fixed, rejected-allograft of a human liver
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.