missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis E Virus Capsid Mouse anti-Virus, Clone: EB2.2.G10.B4.A10, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Hepatitis E Virus Capsid Monoclonal antibody specifically detects Hepatitis E Virus Capsid in Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Hepatitis E Virus Capsid |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | EB2.2.G10.B4.A10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | Phosphate Buffered Saline, pH 7.4, Filter Sterile |
| Gene Alias | ORF2, pORF2, Secreted protein ORF2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?