missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis D virus Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5143460
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Hepatitis D virus Polyclonal antibody specifically detects Hepatitis D virus in Virus samples. It is validated for Western Blot.Specifications
Hepatitis D virus | |
Polyclonal | |
Unconjugated | |
HDV; Hepatitis Delta Virus | |
Full length Hepatitis D virus Small delta recombinant protein (Hepatitis delta virus genotype I (isolate D380)) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.94 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |