missing translation for 'onlineSavingsMsg'
Learn More

Hepatitis D Virus Large/Small Delta Antibody (HL1055), Novus Biologicals™

Product Code. 18334664 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334664 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334664 Supplier Novus Biologicals Supplier No. NBP313704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hepatitis D Virus Large/Small Delta Monoclonal antibody specifically detects Hepatitis D Virus Large/Small Delta in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis D Virus Large/Small Delta
Applications Western Blot
Classification Monoclonal
Clone HL1055
Conjugate Unconjugated
Formulation PBS
Gene Alias Large delta antigen, Large hepatitis delta antigen, L-HDAg, p24, p27, sHD, S-HDAg, Small delta antigen
Host Species Rabbit
Immunogen Full length Hepatitis D virus Large data + Small delta recombinant protein. (Hepatitis delta virus genotype I (isolate D380))
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.