missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hepatitis C Virus NS5b Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111974
This item is not returnable.
View return policy
Description
Hepatitis C Virus NS5b Polyclonal Antibody for Western Blot
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications
| Hepatitis C Virus NS5b | |
| Polyclonal | |
| Unconjugated | |
| HCV; HCV NS5; Hepatitis C Virus; HepC; HepC NS5; NICE-3 | |
| Recombinant protein encompassing a sequence within the center region of Hepatitis C virus NS5B protein (HCV 2a (strain JFH-1)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction