missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hepatitis C Virus NS3 Monoclonal Antibody (C44D)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17345
This item is not returnable.
View return policy
Description
The MA1-7345 antibody reacts with Hepatitis C virus NS3 in viral and infected tissue samples. MA1-7345 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7345 antibody was raised against NS3 protein.
Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
Specifications
| Hepatitis C Virus NS3 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| C44D | |
| Unconjugated | |
| HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3 | |
| NS3 protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction