missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Hepatitis C Virus Core Antigen Polyclonal antibody specifically detects Hepatitis C Virus Core Antigen in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Hepatitis C Virus Core Antigen |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | Core protein p19, HCcAg, HCV core antigen, HCV core protein |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of Hepatitis C Virus Core Antigen (HCV 2a (strain JFH-1)). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?