missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93581AF700
This item is not returnable.
View return policy
Description
Hepatitis C Virus Core Antigen Polyclonal antibody specifically detects Hepatitis C Virus Core Antigen in Human samples. It is validated for Western Blot
Specifications
| Hepatitis C Virus Core Antigen | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
| Hepatitis C Virus (HCV) core protein (HCV-C1) | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 951475 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction