missing translation for 'onlineSavingsMsg'
Learn More

Hepatitis C Virus Antibody, DyLight 755, Novus Biologicals™

Product Code. 18425978 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425978 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425978 Supplier Novus Biologicals Supplier No. NBP227586IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hepatitis C Virus Polyclonal antibody specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5
Host Species Rabbit
Immunogen Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core, env, and part of E2/NS1 of HCV.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.