Learn More
Invitrogen™ Hepatitis B Virus X Monoclonal Antibody (3F6-G10)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA181021
Description
A suggested positive control for immunohistochemical applications is liver carcinoma/hepatitis B infected liver. This product does not require protein digestion pre-treatment of paraffin embedded sections. This product does not require antigen retrieval using heat treatment prior to staining of paraffin embedded sections. Mouse anti Hepatitis B-X antibody, clone 3F6-G10 recognizes the HB-X antigen of hepatitis virus.
The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The Hepatitis B X antigen is a 154 amino acid ~17 kDa multifunctional protein involved in the development of liver chirrosis and hepatocellular carcinoma. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
Specifications
| Hepatitis B Virus X | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA and 0.09% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 3F6-G10 | |
| Unconjugated | |
| HBV; HBVgp3; HepB; HepB X | |
| HB-Xag-Protein A Fusion protein. | |
| 250 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.