Learn More
Invitrogen™ Hepatitis B Virus Surface Monoclonal Antibody (7004-2A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514720
Description
MA5-14720 targets Hepatitis B Surface Antigen in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MA5-14720 immunogen is hepatitis B Surface Antigen. Product MA514720 is a smaller package size of MIH9701 (formerly sold as a Seradyn product).
The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
Specifications
| Hepatitis B Virus Surface | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1 mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| 7004-2A11 | |
| Unconjugated | |
| HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
| Hepatitis B Surface Antigen. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.