missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hepatitis B Virus Surface Ad/Ay Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173087
This item is not returnable.
View return policy
Description
The PA1-73087 antibody reacts with Hepatitis B surface antigen Ad/Ay. PA1-73087 has been successfully used in ELISA applications. The PA1-73087 antibody was raised against surface antigen subtypes ad & ay.
The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The HBV surface protein antigens (HBsAg) are comprised of three carboxyl co terminal HBs proteins termed large (LHBs), middle (MHBs) and small (SHBs, also called major) protein. LHBs and MHBs also share the highly hydrophobic, repetitive, membrane spanning S domain. In addition, MHBs has a 55 amino acid region called preS2. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
Specifications
| Hepatitis B Virus Surface Ad/Ay | |
| Polyclonal | |
| HRP | |
| HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
| Surface antigen subtypes ad & ay. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1-2 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction