missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B surface antigen Goat anti-Hepatitis B virus, Polyclonal, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB29850.500uL
This item is not returnable.
View return policy
Description
Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg).
- High specificity
- High reactivity
Specifications
| Hepatitis B surface antigen | |
| Polyclonal | |
| Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg). | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Native purified hepatitis B virus subtypes Ad and Ay. | |
| RUO | |
| Hepatitis B virus | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user. | |
| Goat | |
| 500 μL | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction