missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68095PEATT594
This item is not returnable.
View return policy
Description
Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Hepatic Sinusoidal Endothelial Cells | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| SE-1 | |
| Flow Cytometry | |
| Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells | |
| Rat Hepatic Sinusoidal Endothelial Cells | |
| 0.1 mL | |
| Human, Mouse, Rat | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction