missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68095AF750
This item is not returnable.
View return policy
Description
Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Hepatic Sinusoidal Endothelial Cells | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SE-1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells | |
| Rat Hepatic Sinusoidal Endothelial Cells | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction