missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepassocin/FGL1 Antibody (1016024) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB16142JF585
This item is not returnable.
View return policy
Description
Hepassocin/FGL1 Monoclonal antibody specifically detects Hepassocin/FGL1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| Hepassocin/FGL1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| fibrinogen-like 1, fibrinogen-like protein 1, hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 | |
| Mouse myeloma cell line NS0-derived recombinant human Hepassocin/FGL1, Met1-Ile312, Accession # BAB70690 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 1016024 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 2267 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction