missing translation for 'onlineSavingsMsg'
Learn More

Heparin Sodium Salt, Thermo Scientific Chemicals

Product Code. 15895548 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mU
100 kU
500 kU
Packaging:
Plastic bottle
Unit Size:
1000ku
100ku
500ku
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15895548 500 kU Plastic bottle 500ku
15815558 1 mU Plastic bottle 1000ku
15885548 100 kU Plastic bottle 100ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15895548 Supplier Thermo Scientific Supplier No. J16920.!T
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 9041-08-1

Specifications

Physical Form Powder
Quantity 500 kU
MDL Number MFCD00081689
Packaging Plastic bottle
Chemical Name or Material Heparin Sodium Salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.