missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparin sodium salt, MedChemExpress
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Description
Heparin sodium salt (Sodium heparin) is an anticoagulant which binds reversibly to antithrombin III (ATIII) and greatly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. Heparin sodium salt significantly inhibits exosome-cell interactions.
Specifications
Specifications
| Chemical Name or Material | Heparin sodium salt |
| Physical Form | Solid |
| CAS | 9041-08-1 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | (C14H25NO20S3)n.xNa |
| Synonym | Sodium heparin Sodium heparinate |
| Solubility Information | H2O : ≥ 150 mg/mL |
| SMILES | O[C@H]1[C@H](O[C@H]2[C@H](OS(O)(=O)=O)[C@@H](O)[C@H](OC)[C@H](C(O)=O)O2)[C@@H](COS(O)(=O)=O)O[C@H](OC)[C@@H]1NS(O)(=O)=O.[n].[.xNa] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction