missing translation for 'onlineSavingsMsg'
Learn More

Heparin sodium salt, from porcine intestinal mucosa, IU≥100/mg, Thermo Scientific Chemicals

Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity.

£26.10 - £267.00

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488890
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16198.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11483277
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16198.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11493277
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16198.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Heparin sodium salt is used as a blood anti-coagulant. It is also used as an effective inhibitor of mesenchymal cell proliferation. Further, it binds to antithrombin III, which inhibits coagulation proteases. It is used as a major component of serum free media for preventing cell aggregation. It is also used as an anti-cancer agent in in vitro cancer research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases

Applications

  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies, this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
Specifications

Chemical Identifiers

9041-08-1
MFCD00081689

Specifications

9041-08-1
14,4653
≥100 IU/mg
MFCD00081689
Soluble in water,dimethyl sulfoxide and ethanol.
Heparin sodium salt, from porcine intestinal mucosa
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : MI0850000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only