missing translation for 'onlineSavingsMsg'
Learn More

Heparin, Fisher BioReagents

Product Code. 10429693
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Packaging:
Poly Vial
Unit Size:
100mg
10mg
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10429693 50 mg Poly Vial 50mg
10001413 10 mg Poly Vial 10mg
10639163 100 mg Poly Vial 100mg
11301146 250 mg Poly Vial 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10429693 Supplier Fisher BioReagents Supplier No. BP252450
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Heparin is used as an anticoagulant in biochemical research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Color White
pH 5.0 to 7.5
Loss on Drying ≤5.0%
Packaging Poly Vial
Quantity 50 mg
Merck Index 15, 4688
Formula Weight ≈5000
Vapor Pressure Negligible
Physical Form Powder or Solid
Chemical Name or Material Heparin, From Porcine Intestinal Mucosa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.