missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Heparin Cofactor II Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547466
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Serpin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency.
Specifications
| Heparin Cofactor II | |
| Polyclonal | |
| Unconjugated | |
| Serpind1 | |
| AA985900; AI303446; D22S673; HC2; Hcf2; HCII; HC-II; Heparin cofactor 2; heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; LS2; Protease inhibitor leuserpin-2; rls2var1; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; serpin D1; serpin family D member 1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1; Serpind1; THPH10 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P05546 | |
| Serpind1 | |
| Sf 21-derived recombinant human Serpin D1/Heparin Cofactor II Gly20-Ser499. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction