missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HepaRG™ Tox Medium Supplement (5X)
Click to view available options
Quantity:
67 mL
Unit Size:
Each
Description
HepaRG™ Tox Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Tox Medium Supplement (5x) is specifically used for applications such as short and long term toxicity studies that require maintenance of hepatocyte-specific functions. The HepaRG™ Tox Medium Supplement (5x) is specifically designed for use with HepaRG™ cells.
Easy media preparation, just thaw, mix, and use
Reproducible, and biologically relevant results with HepaRG™ cells
Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Tox Medium Supplement (5x) to 500 ml of William’s Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.
Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).
HepaRG™ is a trademark of BioPredic International.
Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Tox Medium Supplement (5x) to 500 ml of William’s Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.
Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).
HepaRG™ is a trademark of BioPredic International.
Specifications
Specifications
| Content And Storage | –20°C |
| Description | HepaRG™ Tox Medium Supplement, 67mL, 5X Concentration, Frozen, Sterile, Cell Culture and Research Grade, –20°C Storage, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture, Target-Based ADME/Tox Assays |
| Cell Type | HepaRG Cells |
| Concentration | 5 X |
| Form | Frozen |
| Product Type | Hepatocyte Supplement |
| Sterility | Sterile |
| For Use With (Application) | Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture, Target-Based ADME/Tox Assays |
| Purity or Quality Grade | Cell culture grade, research grade |
| Quantity | 67 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction