missing translation for 'onlineSavingsMsg'
Learn More

Heparanase/HPSE Antibody (SR2148), Novus Biologicals™

Rabbit Monoclonal Antibody

£272.00 - £341.00

Specifications

Antigen Heparanase/HPSE
Clone SR2148
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18612004
View Documents
Novus Biologicals
NBP3-21871-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18669684
View Documents
Novus Biologicals
NBP3-21871-50ul
50 μg
£272.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Heparanase/HPSE Monoclonal antibody specifically detects Heparanase/HPSE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

Heparanase/HPSE
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 3.2, endo-glucoronidase, HEP, heparanase, Heparanase-1, Hpa1, HPAheparanase exon 9 and 10 deletion, HPR1, HPSE1heparanase-1, HSE1heparanase exon10-deletion
A synthesized peptide derived from human Heparanase/HPSE (Uniprot #: Q9Y251)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2148
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Pancreatic Stem Cell Markers, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
10855
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.