missing translation for 'onlineSavingsMsg'
Learn More

Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30731811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731811 Supplier Novus Biologicals Supplier No. AF7276MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Polyclonal antibody specifically detects Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias h3-OST-3B, heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived human Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3, Gly60-Asp390, Accession # Q9Y662
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9953
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.