missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89375
This item is not returnable.
View return policy
Description
Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 Polyclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Heparan Sulfate 3-O-Sulfotransferase 1/HS3ST1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:50 | |
| 3OST, 3-OST-1, 3OST1Heparan sulfate 3-O-sulfotransferase 1, EC 2.8.2, EC 2.8.2.23, h3-OST-1, heparan sulfate (glucosamine) 3-O-sulfotransferase 1, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1, heparan sulfate glucosamine 3-O-sulfotransferase 1, heparin-glucosamine 3-O-sulfotransferase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HS3ST1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLRHIHIVDGDRLIRDPFPEIQKVERFLKLSPQINASNFYFNKTKGFYCLRDSGRDRCLHESKGRAHPQVDPKLLNKLHEYFHEPNKKFFELVGR | |
| 0.1 mL | |
| Signal Transduction | |
| 9957 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction