missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEPACAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599999
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HEPACAM.
HEPACAM is involved in regulating cell motility and cell-matrix interactions. HEPACAM may inhibit cell growth through suppression of cell proliferation.
Specifications
| HEPACAM | |
| Polyclonal | |
| Unconjugated | |
| HEPACAM | |
| 2900042E01Rik; GlialCAM; HEPACAM; hepatic and glial cell adhesion molecule; hepatocyte cell adhesion molecule; Hepn1; hypothetical protein FLJ25530; MLC2A; MLC2B; protein hepaCAM; RGD1306811 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 220296, 300517, 72927 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14CZ8, Q640R3 | |
| HEPACAM | |
| A synthesized peptide derived from human HEPACAM(Accession Q14CZ8), corresponding to amino acid residues S331-A381. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction