missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
Brand: Thermo Scientific™ 760175
Code : 51
Additional Details : Weight : 0.22000kg
Description
HEPA Filters
- 99.97% filtering at 0.3μm
- Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
- Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Specifications
Replacement HEPA Filter | |
For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series incubators |