missing translation for 'onlineSavingsMsg'
Learn More

Heneicosanoic acid, 99%

Product Code. 10266830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266830 1 g Glass bottle 1g
10424195 100 mg Glass bottle 100mg
10461631 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10266830 Supplier Thermo Scientific Acros Supplier No. 204690010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2363-71-5 | C21H42O2 | 326.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2363-71-5
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.55
MDL Number MFCD00002805
InChI Key CKDDRHZIAZRDBW-UHFFFAOYSA-N
Synonym heneicosanoic acid, n-heneicosanoic acid, heneicosylic acid, henicosanoate, heneicosanate, heneicosanoate, n-heneicosanoate, heneicosanoicacid, heneicosanic acid, n-heneicosylic acid
PubChem CID 16898
ChEBI CHEBI:39248
IUPAC Name henicosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 74.0°C to 76.0°C
Color White
Boiling Point 170.0°C (0.1 mbar)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)19CO2H
Quantity 1 g
Beilstein 02, I, 179
Solubility Information Solubility in water: .. Other solubilities: soluble in ether, hot acetone and hot hydrocarbons
Formula Weight 326.55
Percent Purity 99%
Physical Form Shiny Fluffy Crystalline Powder
Chemical Name or Material Heneicosanoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.