missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148360
This item is not returnable.
View return policy
Description
HEN2 Polyclonal Antibody for IHC (P)
May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.
Specifications
| HEN2 | |
| Polyclonal | |
| Unconjugated | |
| NHLH2 | |
| 6230401I09Rik; bHLHa34; bHLHa35; class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; HEN1; HEN-1; Hen2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; Nhlh1; Nhlh2; Nscl; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 4808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q02577 | |
| NHLH2 | |
| E. coli-derived Human NHLH2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction