Learn More
Invitrogen™ HEN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114120
Description
Immunogen sequence: AKYRTAHATR ERIRVEAFNL AFAELRKLLP TLPPDKKLSK IEILRLAICY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The helix-loop-helix (HLH) proteins are a family of putative transcription factors, some of which have been shown to play an important role in growth and development of a wide variety of tissues and species. Four members of this family have been clearly implicated in tumorigenesis via their involvement in chromosomal translocations in lymphoid tumors: MYC (MIM 190080), LYL1 (MIM 151440), E2A (MIM 147141), and SCL (MIM 187040).[supplied by OMIM, Nov 2002].
Specifications
| HEN1 | |
| Polyclonal | |
| Unconjugated | |
| NHLH1 | |
| BHLHA35; class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); HEN1; HEN-1; nescient helix loop helix 1; nescient helix-loop-helix 1; Nhlh1; Nscl; NSCL1; NSCL-1; Tal2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4807 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q02575 | |
| NHLH1 | |
| Synthetic peptide directed towards the middle region of human NHLH1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.