missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEN1/HEN2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total HEN1/2.
The helix-loop-helix (HLH) structures are known motifs commonly found in membrane-active and DNA-binding proteins. Helix-loop-helix proteins (HEN1 and HEN2) are DNA-binding proteins that may be involved in cell-type determination in the early nervous system. Studies of expression in normal tissues have demonstrated expression of NSCL-1 and NSCL-2 (genes encoding HEN1 and HEN2) in the developing central and peripheral nervous system, specifically in developing neurons.
Specifications
Specifications
| Antigen | HEN1/HEN2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NHLH2 |
| Gene Accession No. | Q02575, Q02577 |
| Gene Alias | 6230401I09Rik; bHLHa34; bHLHa35; Class A basic helix-loop-helix protein 34; class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; Hen1; HEN-1; HEN2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; NHLH1; Nhlh2; NSCL; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2 |
| Gene Symbols | NHLH1, NHLH2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?