missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563094
This item is not returnable.
View return policy
Description
Immunogen sequence: LSPDQAADSD HPSSAHSDPE SLGGTDTKVL GSVSDLEPVE EAEGDGKGGS RAALYPHPQQ LSREE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.
Specifications
| HEN2 | |
| Polyclonal | |
| Unconjugated | |
| NHLH2 | |
| 6230401I09Rik; bHLHa34; bHLHa35; class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; HEN1; HEN-1; Hen2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; Nhlh1; Nhlh2; Nscl; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02577 | |
| NHLH2 | |
| Recombinant protein corresponding to Human HEN1/HEN2. Recombinant protein control fragment (Product #RP-101772). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion