missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemopexin Rabbit anti-Human, Mouse, Rat, Clone: 1M6O3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16762-100UL
This item is not returnable.
View return policy
Description
Hemopexin Monoclonal antibody specifically detects Hemopexin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Hemopexin | |
| Monoclonal | |
| Unconjugated | |
| beta-1B-glycoprotein, FLJ56652, hemopexin, HX | |
| A synthetic peptide corresponding to a sequence within amino acids 363-462 of human Hemopexin (P02790). | |
| 100 μg | |
| Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
| 3263 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1M6O3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction