missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Hemopexin Antibody Pair [HRP]

Product Code. 18664470 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18664470

Brand: Novus Biologicals™ NBP2793095Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human Hemopexin.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 18.8 pg/mL
Assay Sensitivity 18.8-1200 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human Hemopexin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta-1B-glycoprotein, FLJ56652, hemopexin, HX
Gene ID (Entrez) 3263
Gene Symbol HPX
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.4 mg/mL of rabbit anti-Human Hemopexin polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 1 mg/mL of mouse anti-Human Hemopexin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 48 ng of recombinant Human Hemopexin.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
    Storage Requirements Storage is content dependent.
    Target Hemopexin
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1200 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.