missing translation for 'onlineSavingsMsg'
Learn More

Hemopexin Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30737660 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737660 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737660 Supplier Novus Biologicals Supplier No. AF7007MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Hemopexin Polyclonal antibody specifically detects Hemopexin in Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemopexin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias beta-1B-glycoprotein, FLJ56652, hemopexin, HX
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Hemopexin, Ser24-Gln460 (His30Asn), Accession # Q91X72
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3263
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.