missing translation for 'onlineSavingsMsg'
Learn More

Hemopexin Antibody (3A9-1A9), Novus Biologicals™

Product Code. 18336777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336777 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336777 Supplier Novus Biologicals Supplier No. H00003263M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hemopexin Monoclonal antibody specifically detects Hemopexin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemopexin
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3A9-1A9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05395
Gene Alias beta-1B-glycoprotein, FLJ56652, hemopexin, HX
Host Species Mouse
Immunogen HPX (AAH05395, 1 a.a. ~ 254 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARVLGAPVALGLWSLCWSLAIATPLPPTSAHGNVAEGETKADPDVTERCSDGWSFDATTLDDNGTMLFFKGEFVWKSHKWDRELISERWKNFPSPVDAAFRQGHNSVFLIKGDKVWVYPPEKKEKGYPKLLQDEFPGIPSPLDAAVECHRGECQAEGVLFFQGDREWFWDLATGTMKERSWPAVGNCSSALRWLGRYYCFQGNQFLRFDPVRGEVPPRYPRDVRDYFMPCPGRGHGHRNGTGHGNSICAVAHI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3263
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.